Merck
  • Stereoselective metal-free synthesis of β-amino thioesters with tertiary and quaternary stereogenic centers.
Stereoselective metal-free synthesis of β-amino thioesters with tertiary and quaternary stereogenic centers.
Angewandte Chemie (International ed. in English) (2014-03-20)
Annette Bahlinger, Sven P Fritz, Helma Wennemers
ABSTRACT
β-Amino thioesters are important natural building blocks for the synthesis of numerous bioactive molecules. An organocatalyzed Mannich reaction was developed which provides direct and highly stereoselective access to acyclic β(2)- and β(2,3,3)-amino thioesters with adjacent tertiary and quaternary stereocenters. Mechanistic studies showed that the stereochemical course of the reaction can be controlled by the choice of the substrates. The β-amino thioesters were further functionalized by, for example, stereoselective decarboxylation to access β(2,3)-frameworks. In addition, the value of the β-amino thioesters was shown in coupling-reagent-free peptide synthesis.
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Urea, European Pharmacopoeia (EP) Reference Standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, analytical standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)