Merck
  • Axonal protection by modulation of p62 expression in TNF-induced optic nerve degeneration.
Axonal protection by modulation of p62 expression in TNF-induced optic nerve degeneration.
Neuroscience letters (2014-08-26)
Kaori Kojima, Yasushi Kitaoka, Yasunari Munemasa, Ayano Hirano, Kana Sase, Hitoshi Takagi
ABSTRACT
p62, which is also called sequestosome 1 (SQSTM1), plays a critical role in neuronal cell death. However, the role of p62 in axonal degeneration remains unclear. We evaluated whether the modulation of p62 expression may affect axonal loss in tumor necrosis factor (TNF)-induced optic nerve degeneration. Immunoblot analysis showed that p62 was upregulated in the optic nerve after intravitreal injection of TNF. Treatment with p62 small interfering RNA (siRNA) exerted a partial but significant protective effect against TNF-induced axonal loss. Rapamycin exerted substantial axonal protection after TNF injection. We found that the increase in p62 was significantly inhibited by p62 siRNA. Treatment with rapamycin also significantly inhibited increased p62 protein levels induced by TNF. These results suggest that the upregulation of p62 may be involved in TNF-induced axonal degeneration and that decreased p62 levels may lead to axonal protection.
Sigma-Aldrich
p-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
p-Phenylenediamine, 98% (GC)
Sigma-Aldrich
p-Phenylenediamine, ≥99.0% (GC/NT)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gtf2h1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dctn4
Sigma-Aldrich
MISSION® esiRNA, targeting human DCTN4
Sigma-Aldrich
MISSION® esiRNA, targeting human GTF2H1