Merck
  • Elevated maternal cortisol leads to relative maternal hyperglycemia and increased stillbirth in ovine pregnancy.
Elevated maternal cortisol leads to relative maternal hyperglycemia and increased stillbirth in ovine pregnancy.
American journal of physiology. Regulatory, integrative and comparative physiology (2014-06-13)
Maureen Keller-Wood, Xiaodi Feng, Charles E Wood, Elaine Richards, Russell V Anthony, Geoffrey E Dahl, Sha Tao
ABSTRACT
In normal pregnancy, cortisol increases; however, further pathological increases in cortisol are associated with maternal and fetal morbidities. These experiments were designed to test the hypothesis that increased maternal cortisol would increase maternal glucose concentrations, suppress fetal growth, and impair neonatal glucose homeostasis. Ewes were infused with cortisol (1 mg·kg(-1)·day(-1)) from day 115 of gestation to term; maternal glucose, insulin, ovine placental lactogen, estrone, progesterone, nonesterified free fatty acids (NEFA), β-hydroxybutyrate (BHB), and electrolytes were measured. Infusion of cortisol increased maternal glucose concentration and slowed the glucose disappearance after injection of glucose; maternal infusion of cortisol also increased the incidence of fetal death at or near parturition. The design of the study was altered to terminate the study prior to delivery, and post hoc analysis of the data was performed to test the hypothesis that maternal metabolic factors predict the fetal outcome. In cortisol-infused ewes that had stillborn lambs, plasma insulin was increased relative to control ewes or cortisol-infused ewes with live lambs. Maternal cortisol infusion did not alter maternal food intake or plasma NEFA, BHB, estrone, progesterone or placental lactogen concentrations, and it did not alter fetal body weight, ponderal index, or fetal organ weights. Our study suggests that the adverse effect of elevated maternal cortisol on pregnancy outcome may be related to the effects of cortisol on maternal glucose homeostasis, and that chronic maternal stress or adrenal hypersecretion of cortisol may create fetal pathophysiology paralleling some aspects of maternal gestational diabetes.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Supelco
Hydrocortisone 21-hemisuccinate, analytical standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Hydrocortisone hydrogen succinate, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard