Merck
  • Development and validation of an ultra-performance liquid chromatography method for simultaneous analysis of 20 antihistaminics in dietary supplements.
Development and validation of an ultra-performance liquid chromatography method for simultaneous analysis of 20 antihistaminics in dietary supplements.
Biomedical chromatography : BMC (2014-08-01)
Jung Yeon Kim, Jung-Ah Do, Ji Yeon Choi, Sooyeul Cho, Woo-Seong Kim, Chang-Yong Yoon
ABSTRACT
The purpose of this study was to develop and validate an ultra-performance liquid chromatography method for simultaneous analysis of 20 antihistamines (illegal additives) in dietary supplements. The limits of detection and quantitation of the method ranged from 1.5 to 2.5 µg/mL and from 20.0 to 50.0 µg/mL, respectively. The determination coefficient was >0.999, precisions were 0.2-5.1% (intra-day) and 0.1-8.8% (inter-day), and accuracies were 84.5-111.2% (intra-day) and 91.9-112.0% (inter-day). The mean recoveries of 20 targeted compounds from dietary supplements ranged from 75.4 to 119.3%. The relative standard deviations were <6.6% and complied with established international guidelines. The relative standard deviation of stability was <0.8%. Fifty-two commercially available dietary supplements were evaluated using this method, and were found to have none of the 20 antihistamines in significant abundance.
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Fexofenadine hydrochloride, >98% (HPLC)
Supelco
Diphenhydramine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Terfenadine
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Hydroxyzine dihydrochloride, ≥97%
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
USP
Olopatadine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Clemastine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clemastine fumarate salt, ≥98% (HPLC)
Diphenhydramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Fexofenadine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
USP
Fexofenadine hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Diphenhydramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Terfenadine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Cyproheptadine hydrochloride sesquihydrate, 99%
Sigma-Aldrich
N,N′-Bis(3-methylphenyl)-N,N′-diphenylbenzidine, 99%
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Promethazine hydrochloride