Merck
  • Heparin inhibits burn-induced spleen cell apoptosis by suppressing interleukin-1 expression.
Heparin inhibits burn-induced spleen cell apoptosis by suppressing interleukin-1 expression.
Chinese medical journal (2014-07-06)
Songfeng Zhao, Xiao Zhang, Xiaojian Zhang, Xiuqin Shi, Zujiang Yu, Quancheng Kan
ABSTRACT
Epidermal burn injury may trigger significant apoptosis of the spleen cells, which might be caused by a burn-induced systemic inflammatory reaction. Heparin has been shown to possess anti-inflammatory properties. Interleukin 1 (IL-1) is centrally important among pro-inflammatory cytokines. We hypothesized that heparin might inhibit burn-induced apoptosis in the spleen via suppression of the IL-1 pathway. Burn injury was performed on IL-1 R+/+ ( IL-1 receptor wild-type mouse) and IL-1 R-/- (IL-1 receptor knock-out mouse) mice, and they were then treated with heparin, saline or IL-1 receptor antagonist IL-Ra. Apoptosis, IL-1α and IL-1β expression were assessed in the spleens and serum. Survival curve analysis was further applied to elucidate the mechanism of heparin's protective properties. Burn induced significant apoptosis (sham: 3.6%± 2.1% vs. burn: 28.8%± 5.9%; P < 0.001) and remarkable expression o IL-1α and IL-1β in the mouse spleens and serum. Heparin reduced the burn-induced apoptosis in the spleens (heparin treated: 8.6%± 3.4%, P < 0.005), which could be blocked by IL-1Ra. Heparin markedly decreased both IL-1α and IL-1β expression in the spleens and serum of burned mice. IL-1 R-/- mice demonstrated considerably less apoptosis in the spleens and had a higher survival rate after burns. Heparin did not significantly decrease apoptosis in the spleen and the mortality rate in IL-1 R-/- mice after burns. Heparin inhibits burn-induced apoptosis of the spleen cells by suppressing IL-1 expression in mice.
Sigma-Aldrich
Rat IL-1 α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-1 α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL1-7R ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Human IL-1 α ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-1 α ELISA Kit, for serum, plasma, cell culture supernatant and urine