Merck
  • Increased benefit of interleukin-1 inhibition on vascular function, myocardial deformation, and twisting in patients with coronary artery disease and coexisting rheumatoid arthritis.
Increased benefit of interleukin-1 inhibition on vascular function, myocardial deformation, and twisting in patients with coronary artery disease and coexisting rheumatoid arthritis.
Circulation. Cardiovascular imaging (2014-05-02)
Ignatios Ikonomidis, Stavros Tzortzis, Ioanna Andreadou, Ioannis Paraskevaidis, Chrysoula Katseli, Pelagia Katsimbri, George Pavlidis, John Parissis, Dimitrios Kremastinos, Maria Anastasiou-Nana, John Lekakis
ABSTRACT
We investigated the effects of anakinra, an interleukin-1 receptor antagonist, on coronary and left ventricular function in coronary artery disease (CAD) patients with rheumatoid arthritis. In a double-blind crossover trial, 80 patients with rheumatoid arthritis (60 with CAD and 20 without) were randomized to a single injection of anakinra or placebo and after 48 hours to the alternative treatment. At baseline and 3 hours after treatment, we assessed (1) flow-mediated dilation of brachial artery; (2) coronary flow reserve, ejection fraction, systemic arterial compliance, and resistance by echocardiography; (3) left ventricular global longitudinal and circumferential strain, peak twisting, untwisting velocity by speckle tracking; and (4) interleukin-1β, nitrotyrosine, malondialdehyde, protein carbonyl, and Fas/Fas ligand levels. At baseline, patients with CAD had 3-fold higher interleukin-1β, protein carbonyl, higher nitrotyrosine, malondialdehyde, and Fas/Fas ligand than non-CAD (P<0.05). After anakinra, there was a greater improvement of flow-mediated dilation (57±4% versus 47±5%), coronary flow reserve (37±4% versus 29±2%), arterial compliance (20±18% versus 2±17%), resistance (-11±19% versus 9±21%), longitudinal strain (33±5% versus 18±2%), circumferential strain (22±5% versus 13±5%), peak twisting (30±5% versus 12±5%), untwisting velocity (23±5% versus 13±5%), ejection fraction (12±5% versus 0.5±5%), apoptotic and oxidative markers, and, in particular, of protein carbonyl (35±20% versus 14±9%) in CAD than in non-CAD patients (P<0.01). No changes in the examined markers were observed after placebo. Interleukin-1 inhibition causes a greater improvement in endothelial, coronary aortic function in addition to left ventricular myocardial deformation and twisting in rheumatoid arthritis patients with CAD than in those without. http://www.clinicaltrials.gov. Unique identifier: NCT01566201.
Sigma-Aldrich
Mouse Fas ELISA Kit
Sigma-Aldrich
Human Fas Ligand ELISA Kit, for serum, plasma, and cell culture supernatants
Sigma-Aldrich
Rat Fas Ligand ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human Fas ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat Fas / Tumor Necrosis Factor Receptor Superfamily Member 6 ELISA Kit
Sigma-Aldrich
Mouse Fas L ELISA Kit
Sigma-Aldrich
Human Fas ELISA Kit, for serum, plasma, cell culture supernatant and urine