Merck
  • Molecular interactions between human cartilaginous endplates and nucleus pulposus cells: a preliminary investigation.
Molecular interactions between human cartilaginous endplates and nucleus pulposus cells: a preliminary investigation.
Spine (2014-05-17)
Cornelia Neidlinger-Wilke, Antje Boldt, Christoph Brochhausen, Fabio Galbusera, Claus Carstens, Franz Copf, Markus Schultheiss, Aron Lazary, Marco Brayda-Bruno, Anita Ignatius, Hans-Joachim Wilke
ABSTRACT
Conditioned media (CM) of cartilaginous endplates (CEPs) of intervertebral discs were analyzed in a bioassay with regard to their influence on matrix turnover and inflammatory factors on nucleus pulposus (NP) cells of the same patient. CEP tissue underwent further histological and ultrastructural analysis. To identify possible interactions between the CEP and the disc via molecular factors that may influence disc matrix degradation and to determine degenerative changes of CEP tissue. Impaired endplate perme-ability due to degeneration and calcification is considered to be a key contributor to disc degeneration. An upregulation of metalloproteinases and inflammatory cytokines has been observed in degenerated intervertebral discs. Possibly, the CEP contributes to the regulation of disc matrix degradation via molecular interactions with the disc tissue. CEP and NP cells from the same patients (n = 6) were investigated in a bioassay with regard to their influence on matrix turnover and inflammatory factors. We determined gene expression of NP cells in alginate beads that were exposed to CM of CEP punches (CEP-CM) from the same patients. The CEP-CMs were analyzed by protein array for inflammatory cytokines. Further CEP samples underwent histological (n = 15) and ultrastructural analysis (n = 8) to determine alterations of cell and matrix structure. NP cells exposed to their donor-corresponding CEP-CM significantly upregulated interleukins (IL-6, IL-8) and matrix metalloproteinase (MMP-3, MMP-13) expression, and significantly decreased aggrecan and collagen type 2 expression. Proinflammatory cytokines were identified in the CEP-CM. The occurrence of apoptotic cells and degraded matrix fragments varied strongly between donors. Our results indicate interactions between the CEP and the NP tissue via molecular factors that upregulate matrix degrading enzymes and inflammatory cytokines and thereby influence the pathophysiology of disc degeneration. Ongoing investigations will further identify the regulative role of potential molecular factors that are responsible for these degenerative alterations. N/A.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
USP
1-Propanol, United States Pharmacopeia (USP) Reference Standard
Supelco
1-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%