Merck
  • Early-onset inflammatory bowel disease and common variable immunodeficiency-like disease caused by IL-21 deficiency.
Early-onset inflammatory bowel disease and common variable immunodeficiency-like disease caused by IL-21 deficiency.
The Journal of allergy and clinical immunology (2014-04-22)
Elisabeth Salzer, Aydan Kansu, Heiko Sic, Peter Májek, Aydan Ikincioğullari, Figen E Dogu, Nina Kathrin Prengemann, Elisangela Santos-Valente, Winfried F Pickl, Ivan Bilic, Sol A Ban, Zarife Kuloğlu, Arzu Meltem Demir, Arzu Ensari, Jacques Colinge, Marta Rizzi, Hermann Eibel, Kaan Boztug
ABSTRACT
Alterations of immune homeostasis in the gut can result in development of inflammatory bowel disease (IBD). Recently, Mendelian forms of IBD have been discovered, as exemplified by deficiency of IL-10 or its receptor subunits. In addition, other types of primary immunodeficiency disorders might be associated with intestinal inflammation as one of their leading clinical presentations. We investigated a large consanguineous family with 3 children who presented with early-onset IBD within the first year of life, leading to death in infancy in 2 of them. Homozygosity mapping combined with exome sequencing was performed to identify the molecular cause of the disorder. Functional experiments were performed to assess the effect of IL-21 on the immune system. A homozygous mutation in IL21 was discovered that showed perfect segregation with the disease. Deficiency of IL-21 resulted in reduced numbers of circulating CD19(+) B cells, including IgM(+) naive and class-switched IgG memory B cells, with a concomitant increase in transitional B-cell numbers. In vitro assays demonstrated that mutant IL-21(Leu49Pro) did not induce signal transducer and activator of transcription 3 phosphorylation and immunoglobulin class-switch recombination. Our study uncovers IL-21 deficiency as a novel cause of early-onset IBD in human subjects accompanied by defects in B-cell development similar to those found in patients with common variable immunodeficiency. IBD might mask an underlying primary immunodeficiency, as illustrated here with IL-21 deficiency.
Sigma-Aldrich
Interleukin-21 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
IL-21 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-21 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture