Merck
  • The carnitine transporter SLC22A5 is not a general drug transporter, but it efficiently translocates mildronate.
The carnitine transporter SLC22A5 is not a general drug transporter, but it efficiently translocates mildronate.
Drug metabolism and disposition: the biological fate of chemicals (2008-11-05)
Silke Grigat, Christian Fork, Markus Bach, Stefan Golz, Andreas Geerts, Edgar Schömig, Dirk Gründemann
ABSTRACT
In addition to its function as carnitine transporter, novel organic cation transporter type 2 (OCTN2; human gene symbol SLC22A5) is widely recognized as a transporter of drugs. This notion is based on several reports of direct measurement of drug accumulation. However, a rigorous, comparative, and comprehensive analysis of transport efficiency of OCTN2 has not been available so far. In the present study, OCTN2 orthologs from human, rat, and chicken were expressed in 293 cells using an inducible expression system. Uptake of trans-4-[4-(dimethylamino)styryl]-1-methylpyridinium iodide (ASP(+)), cephaloridine, ergothioneine, gabapentin, mildronate, pyrilamine, quinidine, spironolactone, tetraethylammonium, verapamil, and vigabatrin was determined by liquid chromatography/mass spectrometry. For reference, uptake of carnitine was measured in parallel. Our results indicate that OCTN2-mediated uptake of drugs was not significantly different from zero or, with tetraethylammonium and ergothioneine, was minute relative to carnitine. The carnitine congener mildronate, by contrast, was transported very efficiently. Thus, OCTN2 is not a general drug transporter but a highly specific carrier for carnitine and closely related molecules. Transport parameters (cellular accumulation, transporter affinity, sodium dependence) were similar for mildronate and carnitine. Efficiency of transport of mildronate was even higher than that of carnitine. Hence, our results establish that OCTN2 is a key target of the cardioprotective agent mildronate because it controls, as integral protein of the plasma membrane, cellular entry of mildronate and enables efficient access to intracellular targets. The highest levels of human OCTN2 mRNA were detected by real-time reverse transcription-polymerase chain reaction in kidney, ileum, breast, small intestine, skeletal muscle, and ovary but also in some heart and central nervous system tissues.
Sigma-Aldrich
Spironolactone, 97.0-103.0%
Sigma-Aldrich
trans-4-[4-(Dimethylamino)styryl]-1-methylpyridinium iodide, Dye content 98 %
Sigma-Aldrich
Gabapentin, solid