Merck
  • EdU incorporation is an alternative non-radioactive assay to [(3)H]thymidine uptake for in vitro measurement of mice T-cell proliferations.
EdU incorporation is an alternative non-radioactive assay to [(3)H]thymidine uptake for in vitro measurement of mice T-cell proliferations.
Journal of immunological methods (2009-08-04)
Yongmao Yu, Alka Arora, Weixian Min, Chaim M Roifman, Eyal Grunebaum
ABSTRACT
T lymphocyte proliferations can be measured by [(3)H]thymidine incorporation. However, many labs avoid this technique because of the need to use radioactive substrates. In addition, [(3)H]thymidine incorporation method does not permit simultaneous characterization of the proliferating cells. We developed the 5-ethynyl-2'-deoxyuridine (EdU) and Cu(I)-catalyzed cycloaddition "click" reaction assay to measure T-cell responses by flow cytometry. Spleen cells from normal, immune-deficient purine nucleoside phosphorylase (PNP) defective (PNP-/-) mice or PNP-/- mice with partial immune reconstitution were stimulated with anti-CD3 antibodies. The correlation (r) between [(3)H]thymidine and EdU incorporations into stimulated T cells was measured and the stimulation index (SI), the ratio between stimulated and non-stimulated cells, was calculated. Flow cytometry was used to characterize the proliferating cells. EdU and [(3)H]thymidine incorporation into normal spleen cells were strongly correlated (r=0.89). Following stimulation, EdU incorporation into spleen cells from normal and immune-reconstituted PNP-/- mice was significantly increased compared to PNP-/- immune-deficient mice. Immune-deficient PNP-/- mice had increased [(3)H]thymidine and EdU incorporation into non-stimulated spleen cells, indicative of spontaneous proliferation. Analysis of EdU incorporation showed that the increased proliferation was due primarily to cells expressing CD3, CD4 and IgM. EdU-Click technology accurately measures proliferation of murine T lymphocyte and can be used as an alternative to [(3)H]thymidine assays. The EdU-Click technology also allows identification of proliferating cells.
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for HTS, sufficient for 200 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for HTS, sufficient for 2000 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for IM IV, sufficient for 100 assays, Kit size M
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for IM, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for IM IV, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for IM, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for IM, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for IM IV, sufficient for 100 assays, Kit size L
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for FC, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for FC, sufficient for 50 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for HTS, sufficient for 400 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for HTS, sufficient for 200 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for FC, sufficient for 100 assays
Sigma-Aldrich
EdU HTS Kit 594, sufficient for 200 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for IM IV, sufficient for 100 assays, Kit size L
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for FC, sufficient for 50 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for IM IV, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for FC, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for IM IV, sufficient for 100 assays, Kit size S
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for FC, sufficient for 50 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for IM IV, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for IM IV size M, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for IM IV, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for FC, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for FC, sufficient for 50 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for IM IV size M, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for IM IV size M, sufficient for 100 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for IM IV, sufficient for 100 assays, Kit size M
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for IM, sufficient for 100 assays