Merck
  • Monitoring of odor compounds produced by solid waste treatment plants with diffusive samplers.
Monitoring of odor compounds produced by solid waste treatment plants with diffusive samplers.
Waste management (New York, N.Y.) (2006-05-23)
P Bruno, M Caselli, G de Gennaro, M Solito, M Tutino
ABSTRACT
Nuisance caused by odors is one of the most important problems for waste management plants. To control an odor nuisance, it must first be quantified. The analytical difficulties in odor measurements are related to the high number of volatile components (belonging to several chemical classes), above all when the concentration is lower than the detection limit of the technique used for the measurement. In this work, 2-butanone, alpha-pinene, tetrachloroethylene, dimethyldisulfide, beta-pinene, limonene, phenol and benzoic acid are determined, because they are representative of some important classes of compounds with higher odor impact. The compounds are sampled with thermal desorbable radial diffusive samplers Radiello containing Tenax cartridges. The analytical repeatability and the complete thermal desorption of the cartridges were verified for each odor compound; the relative standard deviations for repeated samples and the recovery percentage were, respectively, less than 7% and about 97% for all compounds. The measurements of the linearity of sampling showed no systematic difference according to the collection period. The comparison between the odor threshold and the limit of detection demonstrated that this method is reliable for the recognition and quantification of odor compounds, allowing Public Administration to impose legal limits and the control agencies to verify them.
Product Description
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello BTEX/VOC Starter Kit, Thermal Desorption
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello On-Field Thermometer, Reader, pk of 1
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello Filtration Kit, pk of 20
radiello Hydrogen Sulfide Starter Kit
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Barcode Labels, pk of 198
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling Phenolic Compounds (thermal desorption), matrix SS net (100 mesh, 4.8 mm diam.), Tenax-TA, pk of 20
radiello Outdoor Shelter, pk of 10