Merck
  • Regulation of gene expression by cell-to-cell communication: acyl-homoserine lactone quorum sensing.
Regulation of gene expression by cell-to-cell communication: acyl-homoserine lactone quorum sensing.
Annual review of genetics (2001-11-09)
C Fuqua, M R Parsek, E P Greenberg
ABSTRACT
Quorum sensing is an example of community behavior prevalent among diverse bacterial species. The term "quorum sensing" describes the ability of a microorganism to perceive and respond to microbial population density, usually relying on the production and subsequent response to diffusible signal molecules. A significant number of gram-negative bacteria produce acylated homoserine lactones (acyl-HSLs) as signal molecules that function in quorum sensing. Bacteria that produce acyl-HSLs can respond to the local concentration of the signaling molecules, and high population densities foster the accumulation of inducing levels of acyl-HSLs. Depending upon the bacterial species, the physiological processes regulated by quorum sensing are extremely diverse, ranging from bioluminescence to swarming motility. Acyl-HSL quorum sensing has become a paradigm for intercellular signaling mechanisms. A flurry of research over the past decade has led to significant understanding of many aspects of quorum sensing including the synthesis of acyl-HSLs, the receptors that recognize the acyl-HSL signal and transduce this information to the level of gene expression, and the interaction of these receptors with the transcriptional machinery. Recent studies have begun to integrate acyl-HSL quorum sensing into global regulatory networks and establish its role in developing and maintaining the structure of bacterial communities.
Sigma-Aldrich
N-Heptanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Dodecanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-(3-Hydroxydecanoyl)-DL-homoserine lactone
Sigma-Aldrich
N-(3-Hydroxyoctanoyl)-DL-homoserine lactone, ≥95% (sum of isomers, HPLC)
Sigma-Aldrich
N-(3-Hydroxydodecanoyl)-DL-homoserine lactone, ≥97% (HPLC)
Sigma-Aldrich
N-Decanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-(3-Hydroxytetradecanoyl)-DL-homoserine lactone, ≥96% (HPLC), carbon 64.7-67.3 %
Sigma-Aldrich
N-Octanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)