Merck
  • Alterations of neocortical development and maturation in autism: insight from valproic acid exposure and animal models of autism.
Alterations of neocortical development and maturation in autism: insight from valproic acid exposure and animal models of autism.
Neurotoxicology and teratology (2012-09-13)
Taylor Chomiak, Bin Hu
ABSTRACT
Autism spectrum disorder (ASD) is a behaviourally defined brain disorder affecting approximately 1 in 88 children. Many pathological studies have shown that ASD is frequently associated with grey and white matter changes that can be described by their deviations from the normal trajectory of cortical maturation. For example, during the early (i.e. <2 years) postnatal period there is marked and selective tissue overgrowth in the higher-order temporal and frontal networks involved in emotional, social, and communication functions. In this focused review we first summarize some basic principles of neocortical neural organization and how they are disrupted in ASD. We will then highlight some of the potential mechanisms by which the normal developmental trajectory and organization of neocortical networks can be altered based on animal studies of valproic acid, a teratogen widely used in animal models of ASD. We argue that the trajectory of postnatal cerebral neocortex development may be influenced by several cellular and molecular mechanisms that may all converge to produce a neuropathology characterized by premature or accelerated neuronal growth.
Product Description
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentanoic acid
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®