Merck
Tungsten toxicity.
Chemico-biological interactions (2011-12-21)
Mark L Witten, Paul R Sheppard, Brandon L Witten
ABSTRACT
There is emerging evidence that tungsten has toxic health effects. We summarize the recent tungsten toxicity research in this short review. Tungsten is widely used in many commercial and military applications because it has the second highest melting temperature of any element. Consequently, it is important to elucidate the potential health effects of tungsten.
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Tungsten, rod, 200mm, diameter 3.0mm, centerless ground, 99.95%
Tungsten, rod, 100mm, diameter 2.0mm, centerless ground, 99.95%
Tungsten, wire reel, 10m, diameter 0.01mm, hard, 99.95%
Tungsten, wire reel, 10m, diameter 0.025mm, clean, 99.95%
Tungsten, wire reel, 5m, diameter 0.005mm, gold plated, 99.95%
Tungsten, wire reel, 25m, diameter 0.15mm, clean, 99.95%
Tungsten, wire reel, 25m, diameter 0.2mm, clean, 99.95%
Tungsten, wire reel, 5m, diameter 0.15mm, clean, 99.95%
Tungsten, wire, straight, 100mm, diameter 0.25mm, 99.9+%
Tungsten, wire reel, 500m, diameter 0.1mm, clean, 99.95%
Tungsten, wire reel, 5m, diameter 0.1mm, clean, 99.95%
Tungsten, wire, straight, 300mm, diameter 0.125mm, 99.9+%
Tungsten, wire, straight, 300mm, diameter 0.15mm, 99.9+%
Tungsten, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 5m coil, thickness 0.025mm, coil width 1mm, as rolled, 99.95%
Tungsten, wire reel, 10m, diameter 0.08mm, gold plated, 99.95%
Tungsten, wire reel, 25m, diameter 0.025mm, clean, 99.95%
Tungsten, wire reel, 25m, diameter 0.04mm, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.125mm, clean, 99.95%
Tungsten, wire reel, 25m, diameter 0.02mm, gold plated, 99.95%
Tungsten, wire reel, 10m, diameter 0.38mm, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.004mm, hard, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.3mm, clean, 99.95%
Tungsten, wire reel, 100m, diameter 0.5mm, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.2mm, clean, 99.95%
Tungsten, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.95%
Tungsten, mesh, 50x50mm, nominal aperture 0.2mm, wire diameter 0.05mm, 100 wires/inch, open area 64%, plain weave mesh, 99.95%
Tungsten, mesh, 100x100mm, nominal aperture 0.2mm, wire diameter 0.05mm, 100 wires/inch, open area 64%, plain weave mesh, 99.95%