Merck
  • Metals and health: a clinical toxicological perspective on tungsten and review of the literature.
Metals and health: a clinical toxicological perspective on tungsten and review of the literature.
Military medicine (2007-10-17)
Gijsbert B van der Voet, Todor I Todorov, Jose A Centeno, Wayne Jonas, John Ives, Florabel G Mullick
ABSTRACT
Tungsten and tungsten compounds are considered toxicologically relatively safe. Concern regarding the potential health and environmental effects of depleted uranium and lead in military applications has lead many countries to explore the possibility of applying toxicologically safer metals. Heavy metal tungsten alloy-based munitions have been therefore introduced as a replacement in munitions and as kinetic energy penetrators. Although the toxicological profiles of all these metals are well known, their internalization as embedded shrapnel may be considered a new route for long-term exposure. Studies in experimental animals and cell culture indicate that pellets based on heavy metal tungsten alloy possess carcinogenic potential previously unseen for depleted uranium and/or lead. Other metals in the tungsten alloy such as nickel or cobalt may contribute to such a risk. Accordingly, the long-term tungsten-related health risk is reason for concern. This article reviews toxicological and clinical literature and provides new perspectives on tungsten and tungsten-based alloys.
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Tungsten, rod, 200mm, diameter 3.0mm, centerless ground, 99.95%
Tungsten, rod, 100mm, diameter 2.0mm, centerless ground, 99.95%
Tungsten, wire reel, 10m, diameter 0.01mm, hard, 99.95%
Tungsten, wire reel, 10m, diameter 0.025mm, clean, 99.95%
Tungsten, wire reel, 5m, diameter 0.005mm, gold plated, 99.95%
Tungsten, wire reel, 25m, diameter 0.15mm, clean, 99.95%
Tungsten, wire reel, 25m, diameter 0.2mm, clean, 99.95%
Tungsten, wire reel, 5m, diameter 0.15mm, clean, 99.95%
Tungsten, wire, straight, 100mm, diameter 0.25mm, 99.9+%
Tungsten, wire reel, 500m, diameter 0.1mm, clean, 99.95%
Tungsten, wire reel, 5m, diameter 0.1mm, clean, 99.95%
Tungsten, wire, straight, 300mm, diameter 0.125mm, 99.9+%
Tungsten, wire, straight, 300mm, diameter 0.15mm, 99.9+%
Tungsten, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 5m coil, thickness 0.025mm, coil width 1mm, as rolled, 99.95%
Tungsten, wire reel, 10m, diameter 0.08mm, gold plated, 99.95%
Tungsten, wire reel, 25m, diameter 0.025mm, clean, 99.95%
Tungsten, wire reel, 25m, diameter 0.04mm, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.125mm, clean, 99.95%
Tungsten, wire reel, 25m, diameter 0.02mm, gold plated, 99.95%
Tungsten, wire reel, 10m, diameter 0.38mm, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.004mm, hard, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.3mm, clean, 99.95%
Tungsten, wire reel, 100m, diameter 0.5mm, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.2mm, clean, 99.95%
Tungsten, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.95%
Tungsten, mesh, 50x50mm, nominal aperture 0.2mm, wire diameter 0.05mm, 100 wires/inch, open area 64%, plain weave mesh, 99.95%
Tungsten, mesh, 100x100mm, nominal aperture 0.2mm, wire diameter 0.05mm, 100 wires/inch, open area 64%, plain weave mesh, 99.95%