Merck
Calcipotriol/betamethasone dipropionate for the treatment of psoriasis.
Expert opinion on pharmacotherapy (2006-03-24)
A Charakida, O Dadzie, F Teixeira, M Charakida, G Evangelou, A C Chu
ABSTRACT
The two-compound product calcipotriol/betamethasone dipropionate is arising as a first-line treatment for mild-to-moderate plaque psoriasis. Its beneficial action is attributed to the synergistic effect of its components on keratinocyte proliferation and differentiation, and on inflammation. The good tolerability of the two-compound product is thought to be due to the anti-inflammatory effect of betamethasone. Evidence from short-term (4-12 weeks) and long-term use (> 1 year) has shown a good safety profile. Areas such as the face or skin folds, which are sensitive to the components of the combination, should be avoided. Finally, it is unsuitable for use in unstable psoriasis, in which potent steroids may lead to an increased inflammatory response.
Product Description
Betamethasone dipropionate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Betamethasone, VETRANAL®, analytical standard
Betamethasone dipropionate, European Pharmacopoeia (EP) Reference Standard
Betamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betamethasone, ≥98%
Sigma-Aldrich
Betamethasone 17,21-dipropionate
USP
Betamethasone dipropionate, United States Pharmacopeia (USP) Reference Standard
Supelco
Betamethasone Dipropionate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material