Merck
  • Phenylalanine ammonia lyase, enzyme substitution therapy for phenylketonuria, where are we now?
Phenylalanine ammonia lyase, enzyme substitution therapy for phenylketonuria, where are we now?
Molecular genetics and metabolism (2005-09-17)
Christineh N Sarkissian, Alejandra Gámez
ABSTRACT
Phenylketonuria (PKU) is an autosomal recessive genetic disorder in which mutations in the phenylalanine-4-hydroxylase (PAH) gene result in an inactive enzyme (PAH, EC 1.14.16.1). The effect is an inability to metabolize phenylalanine (Phe), translating into elevated levels of Phe in the bloodstream (hyperphenylalaninemia). If therapy is not implemented at birth, mental retardation can occur. PKU patients respond to treatment with a low-phenylalanine diet, but compliance with the diet is difficult, therefore the development of alternative treatments is desirable. Enzyme substitution therapy with a recombinant phenylalanine ammonia lyase (PAL) is currently being explored. This enzyme converts Phe to the harmless metabolites, trans-cinnamic acid and trace ammonia. Taken orally and when non-absorbable and protected, PAL lowers plasma Phe in mutant hyperphenylalaninemic mouse models. Subcutaneous administration of PAL results in more substantial lowering of plasma and significant reduction in brain Phe levels, however the metabolic effect is not sustained following repeated injections due to an immune response. We have chemically modified PAL by pegylation to produce a protected form of PAL that possesses better specific activity, prolonged half-life, and reduced immunogenicity in vivo. Subcutaneous administration of pegylated molecules to PKU mice has the desired metabolic response (prolonged reduction in blood Phe levels) with greatly attenuated immunogenicity.
Product Description
USP
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Phenylalanine, European Pharmacopoeia (EP) Reference Standard