Merck
  • Radiolabeled alpha-melanocyte-stimulating hormone analogs for receptor-mediated targeting of melanoma: from tritium to indium.
Radiolabeled alpha-melanocyte-stimulating hormone analogs for receptor-mediated targeting of melanoma: from tritium to indium.
Journal of molecular recognition : JMR (2003-10-03)
Alex N Eberle, Sylvie Froidevaux
ABSTRACT
Following the first synthesis of tritiated alpha-melanocyte-stimulating hormone (alpha-MSH, alpha-melanotropin) in 1974 by Medzihradszky et al., several alpha-MSH analogs were designed containing between 2 and 12 tritium atoms, the latter of which displayed a specific radioactivity of 12.21 GBq/micromol (330 Ci/mmol). Similarly, radioiodinated alpha-MSH analogs of high purity, full biological activity and a specific radioactivity of approximately 140 GBq/micromol were obtained. Although tritiated and radioiodinated alpha-MSH became indispensable tools as tracer molecules for numerous in vitro and in vivo studies, above all for receptor identification and characterization as well as for structure-activity studies, they did not fulfill the criteria required for therapeutic in vivo targeting of metastatic melanoma. Therefore, we recently developed alpha-MSH analogs containing the universal metal chelator 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetic acid (DOTA) in different positions of the molecule. As DOTA can equally well incorporate diagnostic (e.g. (111)In, (67,68)Ga) and therapeutic (e.g. (90)Y, (67)Cu) radionuclides, DOTA-MSH compounds may serve for both melanoma scintigraphy and therapy. The analog DOTA-[betaAla(3), Nle(4), Asp(5), D-Phe(7), Lys(10)]-alpha-MSH(3-10) (DOTA-MSH(OCT)), which contains the metal chelator at its N-terminal end, displayed good in vitro MC1R affinity (IC(50) 9.21 nm). In vivo, [(111)In]DOTA-MSH(OCT) exhibited a favorable biodistribution profile after injection in B16-F1 tumor-bearing mice. The radiopeptide was rapidly cleared from blood through the kidneys and, most importantly, accumulated preferentially in the melanoma lesions. Lung and liver melanoma metastases could be clearly imaged on tissue section autoradiographs 4 h after injection of [(111)In]DOTA-MSH(OCT). A comparative study of [(111)In]DOTA-MSH(OCT) with [(111)In]DOTA-[Nle(4), D-Phe(7)]-alpha-MSH ([(111)In]DOTA-NDP-MSH) demonstrated the superiority of the DOTA-MSH(OCT) peptide, particularly with respect to the amount of radioactivity taken up by non-malignant organs, including bone, the most radiosensitive tissue. These results demonstrate that [(111)In]DOTA-MSH(OCT) specifically targets melanoma metastases and represents a lead compound for the development of therapeutic DOTA-MSH analogs.
Product Description
Indium, foil, 4mm disks, thickness 0.075mm, as rolled, 100%
Indium, foil, 6mm disks, thickness 0.125mm, as rolled, 100%
Indium, wire reel, 1m, diameter 1.8mm, as drawn, 99.999%
Indium, wire reel, 0.2m, diameter 0.75mm, as drawn, 99.999%
Indium, wire reel, 0.2m, diameter 1.8mm, as drawn, 99.999%
Indium, foil, not light tested, 50x50mm, thickness 0.008mm, temporary acrylic support, 99.8%
Indium, wire reel, 5m, diameter 1.5mm, as drawn, 99.999%
Indium, wire reel, 0.5m, diameter 1.8mm, as drawn, 99.999%
Indium, wire reel, 1m, diameter 1.0mm, as drawn, 99.999%
Indium, wire reel, 0.1m, diameter 1.8mm, as drawn, 99.999%
Indium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.999%
Indium, foil, not light tested, 25x25mm, thickness 0.02mm, 99.8%
Indium, foil, not light tested, 50x50mm, thickness 0.009mm, temporary acrylic support, 99.8%
Indium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.8%
Indium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.8%
Indium, wire reel, 1m, diameter 1.5mm, as drawn, 99.999%
Indium, wire reel, 0.5m, diameter 0.75mm, as drawn, 99.999%
Indium, foil, not light tested, 50x50mm, thickness 0.0125mm, temporary acrylic support, 99.8%
Indium, foil, not light tested, 25x25mm, thickness 0.04mm, 99.8%
Indium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.999%
Indium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.8%
Indium, foil, not light tested, 50x50mm, thickness 0.0025mm, temporary acrylic support, 99.8%
Indium, wire reel, 5m, diameter 0.75mm, as drawn, 99.999%
Indium, wire reel, 2m, diameter 0.5mm, as drawn, 99.999%
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis