- Laser facial skin resurfacing: discussion on erbium:YAG and CO2 lasers.
Laser facial skin resurfacing: discussion on erbium:YAG and CO2 lasers.
The Journal of otolaryngology (2000-05-20)
G N Marcells, D A Ellis
PMID10819104
ABSTRACT
There are two lasers for resurfacing. The erbium:YAG laser and the CO2 laser both have different physical and physiologic differences when striking the skin. Therefore, each laser gives a different result. This article reviews the use of the Sharplan Silktouch CO2 laser for 4 years and the erbium:YAG laser for 1 year. In the last year, we have performed 40 laser procedures with the erbium:YAG laser. Discussion ensues regarding the differences and similarities of the two lasers as well as the difficulties and complications.
Product Description
Erbium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Erbium, rod, 50mm, diameter 6.35mm, cast, 99%
Erbium, rod, 100mm, diameter 6.35mm, cast, 99%
Erbium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Erbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Erbium, rod, 100mm, diameter 10.0mm, cast, 99%
Erbium, foil, 50x50mm, thickness 1.0mm, as rolled, 99%
Erbium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Erbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Erbium, rod, 50mm, diameter 10.0mm, cast, 99%
Erbium, rod, 50mm, diameter 2.0mm, cast, 99%