Merck
  • Constant activation of the RAF-MEK-ERK pathway as a diagnostic and therapeutic target in hairy cell leukemia.
Constant activation of the RAF-MEK-ERK pathway as a diagnostic and therapeutic target in hairy cell leukemia.
Haematologica (2013-01-26)
Enrico Tiacci, Gianluca Schiavoni, Maria Paola Martelli, Emanuela Boveri, Roberta Pacini, Alessia Tabarrini, Silvia Zibellini, Alessia Santi, Valentina Pettirossi, Elisabetta Fortini, Stefano Ascani, Luca Arcaini, Giorgio Inghirami, Marco Paulli, Brunangelo Falini
ABSTRACT
The BRAF-V600E mutation defines genetically hairy cell leukemia among B-cell leukemias and lymphomas. In solid tumors, BRAF-V600E is known to aberrantly activate the oncogenic MEK-ERK pathway, and targeted BRAF and/or MEK inhibitors have shown remarkable efficacy in clinical trials in melanoma patients. However, the MEK-ERK pathway status in hairy cell leukemia has not been thoroughly investigated. We assessed phospho-ERK expression in 37 patients with hairy cell leukemia and 44 patients with neoplasms mimicking hairy cell leukemia (40 splenic marginal zone lymphoma, 2 hairy cell leukemia-variant and 2 splenic lymphoma/leukemia unclassifiable) using immunohistochemistry on routine biopsies and/or Western blotting on purified leukemic cells, and correlated the phospho-ERK status with the BRAF-V600E mutation status. Besides confirming the constant presence of BRAF-V600E in all patients with hairy cell leukemia, we observed ubiquitous phospho-ERK expression in this malignancy. Conversely, all 44 cases with neoplasms mimicking hairy cell leukemia were devoid of BRAF-V600E and none expressed phospho-ERK. Furthermore, the two exceptionally rare cases of non-hairy cell leukemia unclassifiable chronic B-cell neoplasms previously reported to be BRAF-V600E(+) on allele-specific polymerase chain reaction lacked phospho-ERK expression as well, suggesting the presence of the mutation in only a small part of the leukemic clone in these cases. In conclusion, our findings support the use of phospho-ERK immunohistochemistry in the differential diagnosis between hairy cell leukemia and its mimics, and establish the MEK-ERK pathway as a rational therapeutic target in this malignancy.
Product Description
Genomiphi Hy Kit, Cytiva 25-6600-20, sufficient for 100 reactions
Genomiphi Hy Kit, Cytiva 25-6600-25, sufficient for 1000 reactions
GenomiPhi HY Kit, Cytiva 25-6600-22, sufficient for 25 reactions