- Direct synthesis of pentafluoroethyl copper from pentafluoropropionate as an economical C2F5 source: application to pentafluoroethylation of arylboronic acids and aryl bromides.
Direct synthesis of pentafluoroethyl copper from pentafluoropropionate as an economical C2F5 source: application to pentafluoroethylation of arylboronic acids and aryl bromides.
Organic letters (2014-06-14)
Hiroki Serizawa, Kohsuke Aikawa, Koichi Mikami
PMID24926803
ABSTRACT
The direct synthesis of pentafluoroethyl copper (CuC2F5) from a cuprate reagent and ethyl pentafluoropropionate as one of the most economical and useful pentafluoroethyl sources was accomplished. The advantages of this method are; all the reagents employed are low-cost and operationally simple, and the CuC2F5 reagent is prepared in virtually quantitative yield. Furthermore, the CuC2F5 reagent prepared was successfully applied to two types of pentafluoroethylations with arylboronic acids and aryl bromides to provide the pentafluoroethylated aromatic products in good-to-excellent yields, including large scale operations.
Product Description
Continuous cast copper (O), BCR®, certified reference material, rod
Copper, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Copper, tube, 500mm, outside diameter 1.0mm, inside diameter 0.24mm, wall thickness 0.38mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 10mm disks, thickness 1.0mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.125mm, hard, 99.95+%
Copper - O.F.H.C., foil, 2m coil, thickness 0.025mm, hard, 99.95+%
Copper, wire reel, 0.2m, diameter 0.1mm, aluminum cladding, outside diameter 0.25mm, 99.9%
Copper - O.F.H.C., foil, 1m coil, thickness 0.1mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 1m coil, thickness 0.1mm, hard, 99.95+%
Copper - O.F.H.C., foil, 4mm disks, thickness 0.006mm, hard, 99.95+%
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Copper - O.F.H.C., foil, 2m coil, thickness 0.05mm, annealed, 99.95+%
Copper, tube, 500mm, outside diameter 4.0mm, inside diameter 3.2mm, wall thickness 0.4mm, as drawn, 99.9%
Copper, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.99+%
Copper - O.F.H.C., foil, 4mm disks, thickness 0.010mm, hard, 99.95+%
Copper, tube, 500mm, outside diameter 5.0mm, inside diameter 4mm, wall thickness 0.5mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 10m coil, thickness 0.1mm, hard, 99.95+%
Copper - O.F.H.C., foil, 10mm disks, thickness 0.7mm, half hard, 99.95+%