Merck
  • The enhancer and promoter landscape of human regulatory and conventional T-cell subpopulations.
The enhancer and promoter landscape of human regulatory and conventional T-cell subpopulations.
Blood (2014-03-29)
Christian Schmidl, Leo Hansmann, Timo Lassmann, Piotr J Balwierz, Hideya Kawaji, Masayoshi Itoh, Jun Kawai, Sayaka Nagao-Sato, Harukazu Suzuki, Reinhard Andreesen, Yoshihide Hayashizaki, Alistair R R Forrest, Piero Carninci, Petra Hoffmann, Matthias Edinger, Michael Rehli
ABSTRACT
CD4(+)CD25(+)FOXP3(+) human regulatory T cells (Tregs) are essential for self-tolerance and immune homeostasis. Here, we describe the promoterome of CD4(+)CD25(high)CD45RA(+) naïve and CD4(+)CD25(high)CD45RA(-) memory Tregs and their CD25(-) conventional T-cell (Tconv) counterparts both before and after in vitro expansion by cap analysis of gene expression (CAGE) adapted to single-molecule sequencing (HeliScopeCAGE). We performed comprehensive comparative digital gene expression analyses and revealed novel transcription start sites, of which several were validated as alternative promoters of known genes. For all in vitro expanded subsets, we additionally generated global maps of poised and active enhancer elements marked by histone H3 lysine 4 monomethylation and histone H3 lysine 27 acetylation, describe their cell type-specific motif signatures, and evaluate the role of candidate transcription factors STAT5, FOXP3, RUNX1, and ETS1 in both Treg- and Tconv-specific enhancer architectures. Network analyses of gene expression data revealed additional candidate transcription factors contributing to cell type specificity and a transcription factor network in Tregs that is dominated by FOXP3 interaction partners and targets. In summary, we provide a comprehensive and easily accessible resource of gene expression and gene regulation in human Treg and Tconv subpopulations.
Product Description
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Lysine acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine, analytical standard
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ribonucleic acid from torula yeast, core, Type II-C
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ribonucleic acid from torula yeast, Type VI
Ribonucleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ribonucleic acid from baker′s yeast (S. cerevisiae)
Sigma-Aldrich
Ribonucleic acid diethylaminoethanol salt, Type IX
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material