Merck
  • Case-only gene-environment interaction between ALAD tagSNPs and occupational lead exposure in prostate cancer.
Case-only gene-environment interaction between ALAD tagSNPs and occupational lead exposure in prostate cancer.
The Prostate (2014-02-07)
Christine Neslund-Dudas, Albert M Levin, Andrew Rundle, Jennifer Beebe-Dimmer, Cathryn H Bock, Nora L Nock, Michelle Jankowski, Indrani Datta, Richard Krajenta, Q Ping Dou, Bharati Mitra, Deliang Tang, Benjamin A Rybicki
ABSTRACT
Black men have historically had higher blood lead levels than white men in the U.S. and have the highest incidence of prostate cancer in the world. Inorganic lead has been classified as a probable human carcinogen. Lead (Pb) inhibits delta-aminolevulinic acid dehydratase (ALAD), a gene recently implicated in other genitourinary cancers. The ALAD enzyme is involved in the second step of heme biosynthesis and is an endogenous inhibitor of the 26S proteasome, a master system for protein degradation and a current target of cancer therapy. Using a case-only study design, we assessed potential gene-environment (G × E) interactions between lifetime occupational Pb exposure and 11 tagSNPs within ALAD in black (N = 260) and white (N = 343) prostate cancer cases. Two ALAD tagSNPs in high linkage disequilibrium showed significant interaction with high Pb exposure among black cases (rs818684 interaction odds ratio or IOR = 2.73, 95% CI 1.43-5.22, P = 0.002; rs818689 IOR = 2.20, 95% CI 1.15-4.21, P = 0.017) and an additional tagSNP, rs2761016, showed G × E interaction with low Pb exposure (IOR = 2.08, 95% CI 1.13-3.84, P = 0.019). Further, the variant allele of rs818684 was associated with a higher Gleason grade in those with high Pb exposure among both blacks (OR 3.96, 95% CI 1.01-15.46, P = 0.048) and whites (OR 2.95, 95% CI 1.18-7.39, P = 0.020). Genetic variation in ALAD may modify associations between Pb and prostate cancer. Additional studies of ALAD, Pb, and prostate cancer are warranted and should include black men. Prostate 74:637-646, 2014. © 2014 Wiley Periodicals, Inc.
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Lead, foil, 2m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 1m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 1m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 0.5m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)
Lead, foil, 10mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.007mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.02mm, 99.99+%
Lead, wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%
Lead, foil, 250x250mm, thickness 3.0mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 50mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 100x100mm, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
Lead, foil, 25x25mm, thickness 3.0mm, as rolled, 99.99+%
Lead, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 5m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, light tested, 100x100mm, thickness 0.025mm, 99.99+%