Merck
  • Perinatal multiple exposure to neurotoxic (lead, methylmercury, ethylmercury, and aluminum) substances and neurodevelopment at six and 24 months of age.
Perinatal multiple exposure to neurotoxic (lead, methylmercury, ethylmercury, and aluminum) substances and neurodevelopment at six and 24 months of age.
Environmental pollution (Barking, Essex : 1987) (2014-02-04)
Rejane C Marques, José V E Bernardi, José G Dórea, Maria de Fatima R Moreira, Olaf Malm
ABSTRACT
We studied neurodevelopment in infants from two communities. Children living in the vicinity of tin-ore kilns and smelters - TOKS; n = 51) were compared to children from a fishing village (Itapuã; n = 45). Mean hair-Hg (HHg) concentrations were significantly higher in Itapuã children which received significantly (p = 0.0000001) less mean ethylmercury (88.6 μg) from Thimerosal-containing vaccines (TCV) than the TOKS children (120 μg). Breast-milk Pb concentrations were significantly higher in the TOKS mothers (p = 0.000017; 10.04 vs. 3.9 μg L(-1)). Bayley mental development index (MDI) and psychomotor development index (PDI) were statistically significant (respectively p < 0.0000001, p = 0.000007) lower for the TOKS children only at 24 months of age. Multivariate regression analysis showed that MDI was negatively affected by breast-milk Pb and by HHg. PDI was positively affected by breastfeeding and negatively affected by ethylmercury. Milestone achievements were negatively affected by breast-milk Pb (age of walking) and by HHg (age of talking).
Product Description
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, wire reel, 5m, diameter 2.0mm, as drawn, 99.999%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, wire reel, 1000m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, microleaf disks, 10mm, thinness 1.0μm, specific density 269.9μg/cm2, removable support, 99.999%
Aluminum, rod, 50mm, diameter 28mm, as drawn, 99.999%
Aluminum, microfoil, disks, 25mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, microleaf disks, 25mm, thinness 0.15μm, specific density 41.28μg/cm2, removable support, 99.999%
Aluminum, rod, 500mm, diameter 5.0mm, as drawn, 99.999%
Aluminum, tube, 0.5m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, rod, 100mm, diameter 12.0mm, as drawn, 99.999%
Aluminum, rod, 500mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, wire reel, 100m, diameter 0.05mm, as drawn, 99.99+%
Aluminum, rod, 200mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, tube, 100mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, sphere, 100pcs, diameter 3.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, rod, 200mm, diameter 20.0mm, as drawn, 99.999%
Aluminum, rod, 100mm, diameter 28mm, as drawn, 99.999%
Aluminum, wire reel, 10m, diameter 0.5mm, as drawn, 99.999%
Aluminum, wire reel, 500m, diameter 0.7mm, as drawn, 99.5%
Aluminum, wire reel, 30m, diameter 0.063mm, annealed, 99.99%