Merck
Dissipative water intrusion in hydrophobic MCM-41 type materials.
Chemical communications (Cambridge, England) (2002-07-18)
T Martin, B Lefevre, D Brunel, A Galarneau, F Di Renzo, F Fajula, P F Gobin, J F Quinson, G Vigier
ABSTRACT
Texture-related features of water intrusion in hydrophobised MCM-41 silicas render these materials especially suitable for energy dissipation in mechanical dampers.
Product Description
Supelco
Silica gel, high-purity grade (7754), pore size 60 Å, 70-230 mesh
Supelco
Silica gel, high-purity grade, with ~15% calcium sulfate and fluorescent indicator, GF254, suitable for thin layer chromatography (TLC)
Supelco
Silica gel, high-purity grade, pore size 60 Å, 70-230 mesh
Supelco
Silica gel, pore size 50-76 Å, suitable for thin layer chromatography (TLC)
Supelco
Silica gel, high-purity grade (Davisil Grade 62), pore size 150 Å, 60-200 mesh
Supelco
Silica gel, high-purity grade (7734), pore size 60 Å, 70-230 mesh
Supelco
Silica gel, high-purity grade (7749), with gypsum binder and fluorescent indicator, suitable for thin layer chromatography (TLC)
Supelco
Silica gel, pore size 60 Å, 60-200 mesh
Supelco
Silica gel spherical, 40-75 μm particle size
Supelco
Silica gel, high-purity grade (Davisil Grade 633), pore size 60 Å, 200-425 mesh particle size
Supelco
Silica gel, high-purity grade (Davisil Grade 923), pore size 30 Å, 100-200 mesh
Supelco
Silica gel, high-purity grade (Davisil Grade 635), pore size 60 Å, 60-100 mesh
Supelco
Silica gel, high-purity grade (Davisil Grade 636), pore size 60 Å, 35-60 mesh particle size
Supelco
Silica gel, high-purity grade (Davisil Grade 646), 35-60 mesh, pore size 150 Å
Supelco
Silica gel, high-purity grade, pore size 60 Å, 200-400 mesh particle size
Supelco
Silica gel, 6-14 mesh
Supelco
Silica gel, high-purity grade (Davisil Grade 12), pore size 22 Å, 28-200 mesh
Supelco
Silica gel, high-purity grade (Davisil Grade 643), pore size 150 Å, 200-425 mesh
Supelco
Silica gel, high-purity grade (9385), pore size 60 Å, 230-400 mesh particle size
Supelco
Silica gel, high-purity grade, pore size 60 Å, 130-270 mesh, for column chromatography