Merck
  • Isospecific polymerization of 1-hexene by C1-symmetric half-metallocene dimethyl complexes of group 4 metals with bidentate N-substituted iminomethylpyrrolyl ligands.
Isospecific polymerization of 1-hexene by C1-symmetric half-metallocene dimethyl complexes of group 4 metals with bidentate N-substituted iminomethylpyrrolyl ligands.
Dalton transactions (Cambridge, England : 2003) (2012-12-14)
Takahiro Yasumoto, Keishi Yamamoto, Hayato Tsurugi, Kazushi Mashima
ABSTRACT
Non-bridged half-metallocene dimethyl complexes of group 4 metals 2a-4a with an N-4-methoxyphenyl(iminomethyl)pyrrolyl ligand 1a were synthesized and characterized by NMR spectroscopy and X-ray analysis. Upon activation with [Ph3C][B(C6F5)4], these complexes became active catalysts for the polymerization of 1-hexene. A series of hafnium complexes with various N-substituents on the imine group of ligands 1b-1g were also prepared and applied as catalysts for 1-hexene polymerization. The activation parameters for the exchange process between the two methyl groups bound to the metal for Cp*MMe2(R-pyr) complexes were estimated by NMR shape analysis at various temperatures. The findings indicated that the transition state of the ligand flipping process might be associated with the isoselectivity of the polymerization reaction.
Sigma-Aldrich
1-Hexene, ≥99%