Merck
  • Surface-enhanced Raman spectroscopy and density functional theory study on 4,4'-bipyridine molecule.
Surface-enhanced Raman spectroscopy and density functional theory study on 4,4'-bipyridine molecule.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-09-22)
Zhiping Zhuang, Jianbo Cheng, Xu Wang, Bing Zhao, Xiaoxia Han, Yulie Luo
ABSTRACT
The molecular geometry and vibrational frequencies of 4,4'-bipyridine (BPE) in the ground state were calculated using density functional theory (DFT) methods (B3LYP) with 6-31++G(d,p) basis set. The optimized geometric bond lengths and bond angles are obtained by DFT employing the hybrid of Beckes non-local three parameter exchange and correlation functional and Lee-Yang-Parr correlation functional. Fourier transform infrared (FT-IR), Fourier transform Raman (FT-Raman) and near-infrared surface-enhanced Raman scattering (NIR-SERS) spectra of BPE on the silver foil substrate have been recorded. All FT-IR, FT-Raman and NIR-SERS band were assigned on the basis of the B3LYP/6-31++G(d,p) method. The vibrational frequencies obtained by DFT(3LYP) are in good agreement with observed results. The NIR-SERS of BPE excited by 1064nm laser line is little difference with that excited by visible laser line. This phenomenon is result to the increase of the contribution of CHEM enhancement effect. Surface selection rules derived from the electromagnetic enhancement model were employed to infer the orientations of BPE on the silver foil substrate surface. Some vibrational frequency which are sensitive to the planar or non-planar structure of BPE, and to the dihedral angle were concluded.
Product Description
Silver, foil, not light tested, 300x300mm, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, 50x50mm, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 0.5m coil, thickness 0.008mm, 99.95+%
Silver, foil, 150x150mm, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95+%
Silver, foil, 25x25mm, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 150x150mm, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 50x50mm, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95+%
Silver, foil, 0.5m coil, thickness 0.003mm, 99.9%
Silver, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Silver, foil, 0.5m coil, thickness 0.025mm, annealed, 99.95+%
Silver, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 25x25mm, thickness 2.0mm, as rolled, 99.95+%
Silver, foil, 0.2m coil, thickness 0.01mm, 99.95+%
Silver, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Silver, foil, 10mm disks, thickness 0.008mm, 99.97%
Silver, foil, 50x50mm, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 10mm disks, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, 25mm disks, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95+%
Silver, foil, 4mm disks, thickness 0.003mm, 99.9%
Silver, foil, 15mm disks, thickness 0.025mm, as rolled, 99.99+%