Merck
  • Room-temperature surface-erosion route to ZnO nanorod arrays and urchin-like assemblies.
Room-temperature surface-erosion route to ZnO nanorod arrays and urchin-like assemblies.
Chemistry (Weinheim an der Bergstrasse, Germany) (2004-10-09)
Zhengquan Li, Yue Ding, Yujie Xiong, Qing Yang, Yi Xie
ABSTRACT
A solution surface-erosion route was successfully employed to produce one-dimensional (1D) ZnO nanostructures. ZnO nanorod arrays and three-dimensional urchin-like assemblies could be selectively obtained with different manipulations. In this process, zinc foil was introduced to an organic solution system and acted both as a reactant and substrate to support the 1D nanostructures obtained. This method, without any template, apparatus, surfactants, or additional heterogenous substrates, has greatly simplified the preparation of oriented 1D ZnO nanostructures. In particular, this simple route could be carried out at room temperature over a period as short as several minutes, thus it could be conveniently transferred to industrial applications. The possible formation mechanism, erosion process, and influence factors were also investigated.
Product Description
Zinc, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 8mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 8mm disks, thickness 0.5mm, as rolled, 99.99+%
Zinc, foil, not light tested, 25x25mm, thickness 0.015mm, 99.9%
Zinc, foil, 6mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, not light tested, 50x50mm, thickness 0.015mm, 99.9%
Zinc, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Zinc, foil, 100x100mm, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 100x100mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 25x25mm, thickness 3mm, as rolled, 99.99+%
Zinc, foil, 25x25mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 15mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 10mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.02mm, 99.9%
Zinc, foil, 50x50mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 1m coil, thickness 0.075mm, as rolled, 99.95+%