Merck
Correcting for electron contamination at dose maximum in photon beams.
Medical physics (1999-05-05)
D W Rogers
ABSTRACT
Data are presented to allow the photon beam quality specifier being used in the new AAPM TG-51 protocol, %dd(10)x, to be extracted from depth-dose data measured with a 1 mm lead foil either 50 cm or 30 cm from the phantom surface. %dd(10)x is the photon component of the percentage depth dose at 10 cm depth for a 10x10 cm2 field on the surface of a phantom at an SSD of 100 cm. The purpose of the foil is to remove the unknown electron contamination from the accelerator head. Monte Carlo calculations are done: (a) to show these electrons are reduced to a negligible level; (b) to calculate the amount of electron contamination from the lead foil at the depth of dose maximum; and (c) to calculate the effect of beam hardening on %dd(10). The analysis extends the earlier work of Li and Rogers [Med. Phys. 21, 791-798 (1994)] which only provided data for the foil at 50 cm. An error in the earlier Monte Carlo simulations is reported and a more convenient method of analyzing and using the data is presented. It is shown that 20% variations in the foil thickness have a negligible effect on the calculated corrections.
Product Description
Lead, foil, 4mm disks, thickness 0.0125mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 4mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 1m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.007mm, 99.99+%
Lead, foil, 0.5m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 1m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 0.5m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)
Lead, foil, 10mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 6mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 500x500mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
Lead, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 5m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 250x250mm, thickness 3.0mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95%