Merck
  • The symbiotic role of Wolbachia in Onchocercidae and its impact on filariasis.
The symbiotic role of Wolbachia in Onchocercidae and its impact on filariasis.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases (2013-02-13)
T Bouchery, E Lefoulon, G Karadjian, A Nieguitsila, C Martin
ABSTRACT
Symbiotic associations between eukaryotes and microorganisms are frequently observed in nature, and range along the continuum between parasitism and mutualism. The genus Wolbachia contains well-known intracellular bacteria of arthropods that induce several reproductive phenotypes that benefit the transmission of the bacteria. Interestingly, Wolbachia bacteria have been found in the Onchocercidae, a family of filarial nematodes, including species that cause human filarial diseases, e.g. lymphatic filariasis and onchocerciasis. The endosymbiont is thought to be mutualistic in the Onchocercidae, and to provide essential metabolites to the filariae. Currently, Wolbachia bacteria are targets of antibiotic therapy with tetracyclines, which have profound effects on the development, viability and fertility of filarial parasites. This overview article presents the Onchocercidae and Wolbachia, and then discusses the origin and the nature of the symbiosis. It highlights the contribution of Wolbachia to the survival of the filariae and to the development of pathology. Finally, the infection control implications for filariases are debated. Potential directions for future research are also discussed.
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material