Merck
  • Solid-phase synthesis and application of double-fluorescent-labeled lipopeptides, containing a CTL-epitope from the measles fusion protein.
Solid-phase synthesis and application of double-fluorescent-labeled lipopeptides, containing a CTL-epitope from the measles fusion protein.
The journal of peptide research : official journal of the American Peptide Society (1999-11-24)
P Hoogerhout, K J Stittelaar, H F Brugghe, J A Timmermans, G J ten Hove, W Jiskoot, J H Hoekman, P J Roholl
ABSTRACT
The mechanism which enables lipopeptides to induce cytotoxicity is not known. By preparing fluorescent-labeled lipopeptides one might unravel the mechanism of their entry into the cell and their intracellular pathway. A method of preparing double-fluorescent-labeled peptides by solid-phase chemistry is described. As model peptides we have chosen analogs of the sequence RRYPDAVYL, which occurs in the measles fusion protein (F438-446) and is an epitope for cytotoxic T lymphocytes. The peptides Pal-K(TMR)KKKRRYPDAVK(FL)L (7) and Pal-K(FL)KKKRRYPDAVK(TMR)L (8), in which Pal is palmitoyl and K(TMR) and K(FL) are Nepsilon-carboxytetramethylrhodamine- and Nepsilon-carboxyfluorescein-labeled lysyl residues, respectively, were prepared and obtained in approximately 30% yield after purification by high-performance liquid chromatography. The fluorescence of fluorescein and tetramethylrhodamine in lipopeptide Pal-K(TMR)KKKRRYPDAVK(FL)L (7) was quenched to 98-99% due to intramolecular interaction of the labels. On incubation with trypsin (i.e. cleavage at the KKKRR-site) the fluorescence of both labels was restored. The intracellular routing of lipopeptide Pal-K(TMR)KKKRRYPDAVK(FL)L was studied with human melanoma cell line, Mel/J, which was transfected with human leukocyte antigen B*2705. It appeared that the double-fluorescent-labeled lipopeptide was able to induce antigen-specific cytotoxicity. Furthermore, preliminary confocal microscopical studies indicated that this lipopeptide is observed intracellularly.
Sigma-Aldrich
6-Carboxy-tetramethylrhodamine N-succinimidyl ester, BioReagent, suitable for fluorescence
Pricing and availability is not currently available.