Merck
  • Rutile-type (Ti,Sn)O₂ nanorods as efficient anode materials toward its lithium storage capabilities.
Rutile-type (Ti,Sn)O₂ nanorods as efficient anode materials toward its lithium storage capabilities.
Nanoscale (2013-02-13)
Yu-Chun Chen, Tai-Feng Hung, Chih-Wei Hu, Ching-Yu Chiang, Chun-Wei Huang, Hui-Chia Su, Ru-Shi Liu, Chih-Hao Lee, Chia-Chin Chang
ABSTRACT
A series of rutile-type (Ti,Sn)O2 solid solutions with nanorod architecture were successfully synthesized in this study by varying their calcination temperatures of tin-modified titanium dioxide (Sn/TiO2) nanocomposites under a nitrogen atmosphere. During the delithiation process, the (Ti,Sn)O2 nanorods obtained at 500 °C delivered a specific capacity of about 300 mA h g(-1) and showed minimal capacity fading even at a high current density of 3 A g(-1).
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent