Merck
  • Energy-gap opening in a Bi110 nanoribbon induced by edge reconstruction.
Energy-gap opening in a Bi110 nanoribbon induced by edge reconstruction.
Physical review letters (2013-02-02)
Jia-Tao Sun, Han Huang, Swee Liang Wong, H-J Gao, Yuan Ping Feng, Andrew Thye Shen Wee
ABSTRACT
Scanning tunnelling microscopy and spectroscopy experiments complemented by first-principles calculations have been conducted to study the electronic structure of 4 monolayer Bi(110) nanoribbons on epitaxial graphene on silicon carbide [4H-SiC(0001)]. In contrast with the semimetal property of elemental bismuth, an energy gap of 0.4 eV is measured at the centre of the Bi(110) nanoribbons. Edge reconstructions, which can facilitate the edge strain energy release, are found to be responsible for the band gap opening. The calculated density of states around the Fermi level are decreased quickly to zero from the terrace edge to the middle of a Bi(110) nanoribbon potentially signifying a spatial metal-to-semiconductor transition. This study opens new avenues for room-temperature bismuth nanoribbon-based electronic devices.
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis