Merck
  • Synthesis of the aminocyclitol units of (-)-hygromycin A and methoxyhygromycin from myo-inositol.
Synthesis of the aminocyclitol units of (-)-hygromycin A and methoxyhygromycin from myo-inositol.
The Journal of organic chemistry (2012-06-06)
Bharat P Gurale, Mysore S Shashidhar, Rajesh G Gonnade
ABSTRACT
Concise and efficient syntheses of the aminocyclitol cores of hygromycin A (HMA) and methoxyhygromycin (MHM) have been achieved starting from readily available myo-inositol. Reductive cleavage of myo-inositol orthoformate to the corresponding 1,3-acetal, stereospecific introduction of the amino group via the azide, and resolution of a racemic cyclitol derivative as its diastereomeric mandelate esters are the key steps in the synthesis. Synthesis of the aminocyclitol core of hygromycin A involved chromatography in half of the total number of steps, and the aminocyclitol core of methoxyhygromycin involved only one chromatography.
Sigma-Aldrich
Hygromycin B solution from Streptomyces hygroscopicus, ≥60% (HPLC), 45-60 mg/mL in H2O
Sigma-Aldrich
Hygromycin B solution from Streptomyces hygroscopicus, ≥60% (HPLC), 45-60 mg/mL in H2O, γ-irradiated
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, suitable for plant cell culture, BioReagent, ≥60% (HPLC), lyophilized powder
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, lyophilized powder