Merck
  • Spotlight on calcipotriene/betamethasone dipropionate in psoriasis vulgaris of the trunk, limbs, and scalp.
Spotlight on calcipotriene/betamethasone dipropionate in psoriasis vulgaris of the trunk, limbs, and scalp.
American journal of clinical dermatology (2011-10-05)
Paul L McCormack
ABSTRACT
Calcipotriene (calcipotriol)/betamethasone dipropionate (calcipotriene 50 μg/g and betamethasone 0.5 mg/g) is a fixed-dose combination of a vitamin D3 analog and a corticosteroid indicated for the once-daily, topical treatment of psoriasis vulgaris of the trunk, limbs, and scalp in adults. Both the ointment (Daivobet®; Dovobet®) and gel (Xamiol®; Daivobet® Gel; Dovobet® Gel) formulations of calcipotriene/betamethasone dipropionate can be used to treat psoriasis vulgaris of the trunk and/or limbs, although the gel formulation was specifically developed for the treatment of scalp psoriasis. This article reviews the efficacy and tolerability of calcipotriene/betamethasone dipropionate in patients with psoriasis vulgaris, as well as summarizing its pharmacologic properties. Calcipotriene/betamethasone dipropionate has low systemic absorption and displays local anti-inflammatory and immunoregulatory properties. It reduces the hyperproliferation of keratinocytes and helps normalize keratinocyte differentiation. In large, well designed clinical trials, calcipotriene/betamethasone dipropionate, either as the ointment or the gel formulation, applied once daily for 4-8 weeks, was more effective than placebo, calcipotriene, or tacalcitol, as well as betamethasone dipropionate in most instances, for the topical, symptomatic treatment of psoriasis vulgaris of the trunk/limbs. Likewise, calcipotriene/betamethasone dipropionate gel applied once daily for 8 weeks was more effective than placebo or either component alone in the topical, symptomatic treatment of psoriasis vulgaris of the scalp. Long-term, once-daily, when required therapy with calcipotriene/betamethasone dipropionate for 52 weeks was more effective than calcipotriene alone for the treatment of scalp psoriasis, and was at least as effective as switching to calcipotriene for 48 weeks after 4 weeks of calcipotriene/betamethasone dipropionate or alternating between calcipotriene/betamethasone dipropionate and calcipotriene every 4 weeks for 52 weeks in the treatment of psoriasis vulgaris of the trunk/limbs. Calcipotriene/betamethasone dipropionate also improved health-related quality of life. Calcipotriene/betamethasone dipropionate was generally well tolerated, with most adverse drug reactions being lesional or perilesional effects of mild or moderate severity. Calcipotriene/betamethasone dipropionate was often associated with fewer lesional/perilesional adverse reactions than calcipotriene or tacalcitol and did not appear to be associated with a higher incidence of corticosteroid-related adverse events during long-term therapy. Pharmacoeconomic analyses predicted calcipotriene/betamethasone dipropionate to be more cost effective than other topical therapies. Thus, calcipotriene/betamethasone dipropionate is an important, effective, once-daily, topical therapy for the symptomatic treatment of psoriasis vulgaris of the trunk, limbs, and scalp.
Product Description
Supelco
Betamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Betamethasone 17,21-dipropionate
Sigma-Aldrich
Betamethasone, ≥98%
USP
Betamethasone dipropionate, United States Pharmacopeia (USP) Reference Standard
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Betamethasone dipropionate, European Pharmacopoeia (EP) Reference Standard
Betamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Betamethasone Dipropionate, Pharmaceutical Secondary Standard; Certified Reference Material
Betamethasone dipropionate for peak identification, European Pharmacopoeia (EP) Reference Standard