Merck
  • Preoperative administration of Sudan III and successful treatment of persistent chylous ascites in a neonate.
Preoperative administration of Sudan III and successful treatment of persistent chylous ascites in a neonate.
Journal of pediatric surgery (2011-05-28)
Lorna Spagnol, Andrea Conforti, Laura Valfrè, Francesco Morini, Pietro Bagolan
ABSTRACT
Congenital chylous ascites is a rare entity, and surgical treatment is confined to selected intractable cases. We report 2 cases of refractory congenital chylous ascites successfully treated with preoperative administration of lipophilic dye (Sudan III) followed by abdominal systematic surgical exploration, cauterization, and fibrin glue application.
Product Description
Supelco
Sudan III, analytical standard
Sigma-Aldrich
Sudan III, BioXtra, certified by the BSC, certified by the Biological Stain Commission
Sigma-Aldrich
Sudan III, Technical grade
Supelco
Sudan III, C.I. 26100