Merck
  • Cadmium (II) and lead (II) transport in a polymer inclusion membrane using tributyl phosphate as mobile carrier and CuFeO(2) as a polarized photo electrode.
Cadmium (II) and lead (II) transport in a polymer inclusion membrane using tributyl phosphate as mobile carrier and CuFeO(2) as a polarized photo electrode.
Journal of hazardous materials (2010-05-19)
Omar Arous, Mourad Amara, Mohamed Trari, Aissa Bouguelia, Hacène Kerdjoudj
ABSTRACT
In this work, a development of polymeric inclusion membranes for the cations separation is reported. The membrane was made up of cellulose triacetate (CTA) with a tributyl phosphate (TBP) incorporated into the polymer as metal ions carrier. The transport of lead (II) and cadmium (II) ions in two membrane systems polymer inclusion membrane (PIM), PIM coupled with photo-chemical electrode using TBP as carrier and 2-nitro phenyl octyl ether (NPOE) or tris ethylhexyl phosphate (TEHP) as plasticizer have been investigated. The membranes: polymer+plasticizer+carrier were synthesized and characterized by FTIR, X-ray diffraction and scanning electron microscopy (SEM). Transports of lead and cadmium have been studied using these systems and the results were compared to commercial cation exchange membrane (CRA). The obtained results showed that for Pb(2+) ion, the concentrations of the strip phase increases using synthesized membranes. The conduction band of the delafossite CuFeO(2) (-1.25 V(SCE)) yields a thermodynamically M(2+) (=Pb(2+), Cd(2+)) photo electrodeposition and speeds up the diffusion process. In all the cases, the potential of the electrode M/M(2+) in the feed compartment increases until a maximum value, reached at approximately 100 min above which it undergoes a diminution.
Sigma-Aldrich
Tributyl phosphate, 97%
Sigma-Aldrich
Tributyl phosphate, puriss., ≥99.0% (GC)
Sigma-Aldrich
Tributyl phosphate, ≥99%
Supelco
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard