Merck
  • Reactivity of the triple ion and separated ion pair of tris(trimethylsilyl)methyllithium with aldehydes: a RINMR study.
Reactivity of the triple ion and separated ion pair of tris(trimethylsilyl)methyllithium with aldehydes: a RINMR study.
Journal of the American Chemical Society (2008-04-19)
Amanda C Jones, Aaron W Sanders, William H Sikorski, Kristin L Jansen, Hans J Reich
ABSTRACT
Low-temperature rapid-injection NMR (RINMR) experiments were performed on tris(trimethylsilyl)methyllithium. In THF/Me2O solutions, the separated ion (1S) reacted faster than can be measured at -130 degrees C with MeI and substituted benzaldehydes (k >/= 2 s -1), whereas the contact ion (1C) dissociated to 1S before reacting. Unexpectedly, the triple ion reacted faster with electron-rich benzaldehydes relative to electron-deficient ones. The addition of HMPA had no effect on the rate of reaction of the triple ion with p-diethylaminobenzaldehyde, and the immediate product of the reaction was the HMPA-solvated separated ion 1S, with the Peterson product forming only slowly. Thus, the aldehyde is catalyzing the dissociation of the triple ion. HMPA greatly decelerated the reaction of 1S (<10 -10), providing an estimate of the Lewis acid activating effect of a THF-solvated lithium cation in an organolithium addition to an aldehyde.
Sigma-Aldrich
Hexamethylphosphoramide, 99%
Sigma-Aldrich
Hexamethylphosphoramide, purum, ≥98.0% (GC)
Sigma-Aldrich
Hexamethylphosphoramide, absolute, over molecular sieve (H2O ≤0.03%), ≥98.0% (GC)