Merck
  • Effect of polyphosphoric acid pre-treatment of titanium on attachment, proliferation, and differentiation of osteoblast-like cells (MC3T3-E1).
Effect of polyphosphoric acid pre-treatment of titanium on attachment, proliferation, and differentiation of osteoblast-like cells (MC3T3-E1).
Clinical oral implants research (2008-01-15)
Kenji Maekawa, Yasuhiro Yoshida, Atsushi Mine, Bart van Meerbeek, Kazuomi Suzuki, Takuo Kuboki
ABSTRACT
To evaluate the effect of polyphosphoric acid (PPA) pre-treatment of titanium (Ti) on the initial attachment, proliferation, and differentiation of mouse osteoblast-like cells (MC3T3-E1). Adsorption of PPA to Ti was achieved by immersing Ti disks (15 mm in diameter) into 0, 1, and 10 wt% PPA and 10 wt% orthophosphoric acid (OPA) for 24 h. On each pre-treated Ti disk, 5.0 x 10(4) MC3T3-E1 cells were seeded, and 1, 3, and 5 h later cell attachment was evaluated. Cell proliferation was also determined 1, 3, and 5 days after cell seed. Cell differentiation was evaluated 5, 10, and 15 days after cell seed using osteoblast marker gene expression. Total RNA was purified from each culture and Type-I collagen, alkaline phosphatase, and osteocalcin mRNA expression levels were measured by real-time reverse transcription polymerase chain reaction. Adsorption of PPA or OPA to Ti significantly accelerated initial cell attachment at every time point (P<0.0001). As with cell attachment, cell proliferation was also accelerated on the PPA-treated Ti disks in a dose-dependent manner at every time point (P<0.0001). However, OPA-treated Ti disks did not enhance the cell proliferation. Regarding osteoblastic differentiation, PPA-treated Ti significantly accelerated the Type-I collagen gene expression at 5 and 10 days after cell seed. Regarding alkaline phosphatase and osteocalcin gene expression, no significant difference was found among the different Ti surface conditions. The accelerated cell behavior following Ti pre-treatment with PPA is a promising new strategy to fabricate bioactive Ti implants.
Sigma-Aldrich
Polyphosphoric acid, ~105% H3PO4 basis
Sigma-Aldrich
Polyphosphoric acid, reagent grade, 115% H3PO4 basis