Merck
  • Immobilization of simulated reducing agent at the surface of SiO2 fillers in dental composite resins.
Immobilization of simulated reducing agent at the surface of SiO2 fillers in dental composite resins.
Dental materials journal (2007-09-25)
Satoki Shibata, Isao Hirata, Yuji Nomura, Kenichi Shirai, Morioki Fujitani, Hideaki Shintani, Masayuki Okazaki
ABSTRACT
To reduce the leachability of reducing agents from composite resins, immobilization of a simulated reducing agent at the surface of SiO2 fillers was examined. SiO2 plates were immersed in 2% 3-aminopropyltriethoxy silane/ethanol solution, and then immersed in dimethyl sulfoxide with 0.25 wt% 4-dimethyl amino benzoic acid (DMABA), 2.0 wt% 1-ethyl-3-(3-dimethylaminopropyl) carbodiimide hydrochloride, and 0.5 wt% N-hydroxysuccinimide. Wide-scan spectrum of X-ray photoelectron spectroscopy did not detect carbon contamination. However, narrow scan detected an O=C-N peak at 399.8 eV, suggesting that DMABA could be immobilized on silane-coupled SiO2 plates. Further, surface plasmon resonance analysis indicated the adsorption of MMA at the surface of reducing agent-immobilized plate.
Sigma-Aldrich
Ethyl 4-(dimethylamino)benzoate, ≥99%
Supelco
Ethyl 4-(dimethylamino)benzoate, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland