Merck
  • The novel and efficient direct synthesis of N,O-acetal compounds using a hypervalent iodine(III) reagent: an improved synthetic method for a key intermediate of discorhabdins.
The novel and efficient direct synthesis of N,O-acetal compounds using a hypervalent iodine(III) reagent: an improved synthetic method for a key intermediate of discorhabdins.
Chemical communications (Cambridge, England) (2005-03-26)
Yu Harayama, Masako Yoshida, Daigo Kamimura, Yasuyuki Kita
ABSTRACT
The use of hypervalent iodine(III) reagents allowed us to develop the novel and efficient direct synthesis of N,O-acetal compounds via the oxidative fragmentation reaction of alpha-amino acids or alpha-amino alcohols; furthermore, we succeeded in developing an improved synthesis of the key intermediate of discorhabdins.
Sigma-Aldrich
DL-Serine methyl ester hydrochloride, 98%
Sigma-Aldrich
L-Serine methyl ester hydrochloride, 98%
Sigma-Aldrich
D-Serine methyl ester hydrochloride, 98%