Merck
  • Olfaction in the Queensland fruit fly, Bactrocera tryoni. II: Response spectra and temporal encoding characteristics of the carbon dioxide receptors.
Olfaction in the Queensland fruit fly, Bactrocera tryoni. II: Response spectra and temporal encoding characteristics of the carbon dioxide receptors.
Journal of chemical ecology (2001-07-27)
C D Hull, B W Cribb
ABSTRACT
Single-unit electrophysiology was used to record the nerve impulses from the carbon dioxide receptors of female Queensland fruit flies, Bactroera tryoni. The receptors responded to stimulation in a phasic-tonic manner and also had a period of inhibition of the nerve impulses after the end of stimulation. at high stimulus intensities. The cell responding to carbon dioxide was presented with a range of environmental odorants and found to respond to methyl butyrate and 2-butanone. The coding characteristics of the carbon dioxide cell and the ability to detect other odorants are discussed, with particular reference to the known behavior of the fly.
Sigma-Aldrich
Methyl butyrate, natural, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl butyrate, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl butyrate, 99%
Pricing and availability is not currently available.
Supelco
Methyl butyrate, analytical standard
Pricing and availability is not currently available.