Merck
  • Study of the formation mechanisms of some volatile compounds during the aging of sweet fortified wines.
Study of the formation mechanisms of some volatile compounds during the aging of sweet fortified wines.
Journal of agricultural and food chemistry (1999-12-20)
I Cutzach, P Chatonnet, D Dubourdieu
ABSTRACT
Sweet fortified wines, traditionally aged under strong oxidation conditions, have a characteristic aroma. An experimental laboratory study investigated the aging of red and white sweet fortified wines under various conditions. The formation of various molecules, previously identified as characteristic of the aroma of this type of wine, was monitored by analysis. The development of these compounds during accelerated aging was affected by oxidation and the color of the wine. Among the molecules studied, sotolon [3-hydroxy-4, 5-dimethyl-2(5H)-furanone] was one of the few molecules present in concentrations above the perception threshold, in both red and white wines. Buildup was strongly affected by the presence of oxygen in white wine subjected to accelerated aging. (Ethoxymethyl)furfural, formed from 5-(hydroxymethyl)furfural, and furfural, derived from sugars, are also involved in the aroma of sweet fortified white wines aged in oxygen-free conditions. The substances most characteristic of accelerated aging of sweet fortified red wines were 5-(hydroxymethyl)furfural, acetylformoin, and hydroxymaltol, the formation of which is affected by oxidation, and dihydromaltol, formed in the absence of oxidation.
Sigma-Aldrich
γ-Octalactone, ≥97%, FCC, FG
Sigma-Aldrich
γ-Octalactone, natural, ≥97%, FG
Sigma-Aldrich
4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, ≥97%, FG
Supelco
4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, analytical standard