Merck
  • Innate immune response to H3N2 and H1N1 influenza virus infection in a human lung organ culture model.
Innate immune response to H3N2 and H1N1 influenza virus infection in a human lung organ culture model.
Virology (2009-11-17)
Wenxin Wu, J Leland Booth, Elizabeth S Duggan, Shuhua Wu, Krupa B Patel, K Mark Coggeshall, Jordan P Metcalf
ABSTRACT
We studied cytokine responses to influenza virus PR8 (H1N1) and Oklahoma/309/06 (OK/06, H3N2) in a novel human lung tissue model. Exposure of the model to influenza virus rapidly activated the mitogen-activated protein kinase signaling (MAPK) pathways ERK, p38 and JNK. In addition, RNase protection assay demonstrated the induction of several cytokine and chemokine mRNAs by virus. This finding was reflected at the translational level as IL-6, MCP-1, MIP-1 alpha/beta, IL-8 and IP-10 proteins were induced as determined by ELISA. Immunohistochemistry for IP-10 and MIP-1 alpha revealed that alveolar epithelial cells and macrophages were the source of these two cytokines. Taken together, both PR8 and OK/06 cause similar induction of cytokines in human lung, although OK/06 is less effective at inducing the chemokines MCP-1 and IL-8. This human organ culture model should thus provide a relevant platform to study the biological responses of human lung to influenza virus infection.
Product Description
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with non-essential amino acids, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s salts, with 2.0 mM L-glutamine, with 20.0 mM HEPES, with non-essential amino acids, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 25mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, with non-essential amino acids, without sodium bicarbonate, dry powder, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, liquid, sterile-filtered, suitable for cell culture