Merck
  • Structural studies of human brain-type creatine kinase complexed with the ADP-Mg2+-NO3- -creatine transition-state analogue complex.
Structural studies of human brain-type creatine kinase complexed with the ADP-Mg2+-NO3- -creatine transition-state analogue complex.
FEBS letters (2008-11-04)
Seoung Min Bong, Jin Ho Moon, Ki Hyun Nam, Ki Seog Lee, Young Min Chi, Kwang Yeon Hwang
ABSTRACT
Creatine kinase is a member of the phosphagen kinase family, which catalyzes the reversible phosphoryl transfer reaction that occurs between ATP and creatine to produce ADP and phosphocreatine. Here, three structural aspects of human-brain-type-creatine-kinase (hBB-CK) were identified by X-ray crystallography: the ligand-free-form at 2.2A; the ADP-Mg2+, nitrate, and creatine complex (transition-state-analogue complex; TSAC); and the ADP-Mg2+-complex at 2.0A. The structures of ligand-bound hBB-CK revealed two different monomeric states in a single homodimer. One monomer is a closed form, either bound to TSAC or the ADP-Mg2+-complex, and the second monomer is an unliganded open form. These structural studies provide a detailed mechanism indicating that the binding of ADP-Mg2+ alone may trigger conformational changes in hBB-CK that were not observed with muscle-type-CK.
Sigma-Aldrich
Creatine Phosphokinase from bovine heart, Type III, salt-free, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein