Merck
"Covalent affinity" purification of ubiquitin-activating enzyme.
The Journal of biological chemistry (1982-03-10)
A Ciechanover, S Elias, H Heller, A Hershko
ABSTRACT
We have previously described an enzyme that activates ubiquitin, the heat-stable polypeptide of the ATP-dependent proteolytic system from reticulocytes (Ciechanover, A., Heller, H., Katz-Etzion, R., and Hershko, A. (1981) Proc. Natl. Acad. Sci. U. S. A. 78, 761-765). It carries out ubiquitin-dependent PPi-ATP and AMP-ATP exchange reactions and binds to the activated polypeptide by a thiolester linkage. We describe here a procedure for the purification of this enzyme by its binding to ubiquitin-Sepharose. Binding of the enzyme to the affinity column requires ATP and Mg2+, and bound enzyme cannot be displaced by high salt but can be eluted by raising the pH, by increased concentrations of a thiol compound, or by the joint supplementation of AMP and pyrophosphate. Another form of the enzyme that cannot carry out AMP-ATP exchange (but catalyzes ubiquitin-dependent PPi-ATP exchange) does not bind to the affinity column. These data indicate that a covalent, possibly thiolester intermediate, is formed between the activating enzyme and Sepharose-bound ubiquitin. It is suggested designating this procedure of enzyme isolation "covalent affinity" chromatography. The purified enzyme has an apparent Mr = 210,000 and appears to be composed of two subunits of Mr = 105, 000. ATP-dependent binding of ubiquitin to the purified enzyme and to its subunit is demonstrated.
Sigma-Aldrich
Ubiquitin−Agarose, saline suspension