Merck
  • Lipopolysaccharide-induced inflammation increases nitric oxide production in taste buds.
Lipopolysaccharide-induced inflammation increases nitric oxide production in taste buds.
Brain, behavior, and immunity (2022-04-22)
Zhizhongbin Wu, Yilin Huang, Weiqing Hu, Liyin Ren, Peihua Jiang, Robert F Margolskee, Hong Wang, Shan Feng
ABSTRACT
Inducible nitric oxide synthase (iNOS) is expressed when cells are induced or stimulated by proinflammatory cytokines and/or bacterial lipopolysaccharide (LPS). iNOS is a downstream gene of the NF-κB pathway. Our previous studies demonstrated that five Nfkb genes are expressed in mouse taste epithelium and taste organoids. However, it is unclear whether activation of the NF-κB pathway could induce iNOS gene expression and increase nitric oxide (NO) production in taste buds. In this study, we investigated the expression of iNOS mRNA and protein after LPS stimulation. Our results showed that a subset of taste bud cells and taste neurons express iNOS proteins after LPS stimulation. In addition, isolated mouse taste epithelium can release NO after exposure to LPS ex vivo. In taste behavioral tests, the NO donor nitroprusside enhanced mouse aversive responses to salty, bitter, and sour taste compounds. The enhanced aversive responses were especially strong for salty taste. In conclusion, our results suggest that iNOS and NO may play a role in the inflammation-associated taste disturbances.
Sigma-Aldrich
Anti-P2X3 Receptor Antibody, pain, serum, Chemicon®