Merck
  • A high-throughput screening identifies MCM chromatin loading inhibitors targeting cells with increased replication origins.
A high-throughput screening identifies MCM chromatin loading inhibitors targeting cells with increased replication origins.
iScience (2024-08-26)
Lucia Falbo, Hervé Técher, Vincenzo Sannino, Michela Robusto, Giovanni Fagà, Federica Pezzimenti, Francesco Romeo, Luca Gabriele Colombo, Stefania Vultaggio, Daniele Fancelli, Silvia Monzani, Valentina Cecatiello, Sebastiano Pasqualato, Mario Varasi, Ciro Mercurio, Vincenzo Costanzo
ABSTRACT
Replication origin assembly is a pivotal step in chromosomal DNA replication. In this process, the ORC complex binds DNA and, together with the CDC6 and CDT1, promotes the loading of the MCM helicase. Chemicals targeting origin assembly might be useful to sensitize highly proliferative cancer cells. However, identifying such compounds is challenging due to the multistage nature of this process. Here, using Xenopus laevis egg extract we set up a high-throughput screening to isolate MCM chromatin loading inhibitors, which led to the identification of NSC-95397 as a powerful inhibitor of replication origin assembly that targets CDC6 protein and promotes its degradation. Using systems developed to test selective drug-induced lethality we show that NSC-95397 triggers cell death both in human cells and Xenopus embryos that have higher proliferative ability. These findings demonstrate the effectiveness of molecules disrupting DNA replication processes in targeting hyperproliferating cells, highlighting their potential as anti-cancer molecules.
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, ≥98% (HPLC), powder
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
Roscovitine, A potent, reversible, and selective inhibitor of Cdks that exhibits about 10-fold greater efficacy towards p34-cdk1 and p33-cdk2 and 20-fold greater efficacy towards p33-cdk5 relative to Olomoucine.
Sigma-Aldrich
Cdc25 Inhibitor IV, NSC 95397, The Cdc25 Inhibitor IV, NSC 95397, also referenced under CAS 93718-83-3, controls the biological activity of Cdc25. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
1-Decanol, ≥98%
Sigma-Aldrich
Spermidine, BioReagent, for molecular biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).