Merck
  • DIRAS3 enhances RNF19B-mediated RAC1 ubiquitination and degradation in non-small-cell lung cancer cells.
DIRAS3 enhances RNF19B-mediated RAC1 ubiquitination and degradation in non-small-cell lung cancer cells.
iScience (2023-07-24)
Yingying Wang, Minli Wei, Min Su, Zhiyuan Du, Jiaxi Dong, Yu Zhang, Yingdi Wu, Xiaopeng Li, Ling Su, Xiangguo Liu
ABSTRACT
Distant metastasis remains the leading cause of high mortality in patients with non-small-cell lung cancer (NSCLC). DIRAS3 is a candidate tumor suppressor protein that is decreased in various tumors. However, the regulatory mechanism of DIRAS3 on metastasis of NSCLC remains unclear. Here, we found that DIRAS3 suppressed the migration of NSCLC cells. Besides, DIRAS3 stimulated the polyubiquitination of RAC1 and suppressed its protein expression. Furthermore, RNF19B, a member of the RBR E3 ubiquitin ligase family, was observed to be the E3 ligase involved in the DIRAS3-induced polyubiquitination of RAC1. DIRAS3 could promote the binding of RAC1 and RNF19B, thus enhancing the degradation of RAC1 by the ubiquitin-proteasome pathway. Finally, the DIRAS3-RNF19B-RAC1 axis was confirmed to be associated with the malignant progression of NSCLC. These findings may be beneficial for developing potential prognostic markers of NSCLC and may provide an effective treatment strategy.
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-RNF19B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution