Merck
  • Deciphering neuronal deficit and protein profile changes in human brain organoids from patients with creatine transporter deficiency.
Deciphering neuronal deficit and protein profile changes in human brain organoids from patients with creatine transporter deficiency.
eLife (2023-10-13)
Léa Broca-Brisson, Rania Harati, Clémence Disdier, Orsolya Mozner, Romane Gaston-Breton, Auriane Maïza, Narciso Costa, Anne-Cécile Guyot, Balazs Sarkadi, Agota Apati, Matthew R Skelton, Lucie Madrange, Frank Yates, Jean Armengaud, Rifat Hamoudi, Aloïse Mabondzo
ABSTRACT
Creatine transporter deficiency (CTD) is an X-linked disease caused by mutations in the SLC6A8 gene. The impaired creatine uptake in the brain results in intellectual disability, behavioral disorders, language delay, and seizures. In this work, we generated human brain organoids from induced pluripotent stem cells of healthy subjects and CTD patients. Brain organoids from CTD donors had reduced creatine uptake compared with those from healthy donors. The expression of neural progenitor cell markers SOX2 and PAX6 was reduced in CTD-derived organoids, while GSK3β, a key regulator of neurogenesis, was up-regulated. Shotgun proteomics combined with integrative bioinformatic and statistical analysis identified changes in the abundance of proteins associated with intellectual disability, epilepsy, and autism. Re-establishment of the expression of a functional SLC6A8 in CTD-derived organoids restored creatine uptake and normalized the expression of SOX2, GSK3β, and other key proteins associated with clinical features of CTD patients. Our brain organoid model opens new avenues for further characterizing the CTD pathophysiology and supports the concept that reinstating creatine levels in patients with CTD could result in therapeutic efficacy.
Sigma-Aldrich
Anti-β-Tubulin III (neuronal) antibody, Mouse monoclonal, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-PSD95 Antibody, clone K28/43, clone K28/43, from mouse
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-PAX6 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL5414, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP2 Antibody, clone 3C19, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells